Ethyl-5-(3-chlorophenyl)[1,2,4]oxadiazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

059034-1G 059034-5G 059034-10G
101916-790EA 378.46 USD
101916-790 101916-792 101916-794
Ethyl-5-(3-chlorophenyl)[1,2,4]oxadiazole-3-carboxylate ≥95%
Ethyl-5-(3-chlorophenyl)[1,2,4]oxadiazole-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₉ClN₂O₃
MDL Number: MFCD10568210
CAS Number: 657424-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR