Ethyl-3-(3-chlorophenyl)[1,2,4]oxadiazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

059033-1G 059033-5G 059033-10G
101916-784EA 378.46 USD
101916-784 101916-786 101916-788
Ethyl-3-(3-chlorophenyl)[1,2,4]oxadiazole-5-carboxylate ≥95%
Ethyl-3-(3-chlorophenyl)[1,2,4]oxadiazole-5-carboxylate

Hazard: Irritant

Formula: C₁₁H₉ClN₂O₃
MDL Number: MFCD02102655
CAS Number: 478030-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR