Ethyl-2-(4-chlorophenoxy)acetoacetate ≥95%

Supplier: Matrix Scientific

059032-1G 059032-5G
101916-780EA 436.92 USD
101916-780 101916-782
Ethyl-2-(4-chlorophenoxy)acetoacetate ≥95%
Ethyl-2-(4-chlorophenoxy)acetoacetate

Hazard: Irritant

Formula: C₁₂H₁₃ClO₄
MDL Number: MFCD10568345
CAS Number: 119138-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR