Ethyl-5-(chloromethyl)-1,2,4-oxadiazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

059031-1G 059031-5G
101916-776EA 438.46 USD
101916-776 101916-778
Ethyl-5-(chloromethyl)-1,2,4-oxadiazole-3-carboxylate ≥95%
Ethyl-5-(chloromethyl)-1,2,4-oxadiazole-3-carboxylate

Hazard: Irritant

Formula: C₆H₇ClN₂O₃
CAS Number: 1009620-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR