Ethyl-1-carbamimidoylpiperidine-4-carboxyateacetate ≥95%

Supplier: Matrix Scientific

059028-1G 059028-5G 059028-10G
101916-762EA 346.15 USD
101916-762 101916-764 101916-766
Ethyl-1-carbamimidoylpiperidine-4-carboxyateacetate ≥95%
Ethyl-1-carbamimidoylpiperidine-4-carboxyateacetate

Hazard: Irritant

Formula: C₁₁H₂₁N₃O₄
MDL Number: MFCD10568351
CAS Number: 1208081-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR