Ethyl-5-bromomethyl-3-methylisoxazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

059027-1G 059027-5G
101916-758EA 346.15 USD
101916-758 101916-760
Ethyl-5-bromomethyl-3-methylisoxazole-4-carboxylate ≥95%
Ethyl-5-bromomethyl-3-methylisoxazole-4-carboxylate

Hazard: Irritant

Formula: C₈H₁₀BrNO₃
MW: 248.08 g/mol
MDL Number: MFCD10568349
CAS Number: 159770-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR