Ethyl-3-amino-4,6-dimethyl-1H-pyrrolo[2,3-b]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

059022-1G 059022-5G 059022-10G
101916-736EA 320 USD
101916-736 101916-738 101916-740
Ethyl-3-amino-4,6-dimethyl-1H-pyrrolo[2,3-b]pyridine-2-carboxylate ≥95%
Ethyl-3-amino-4,6-dimethyl-1H-pyrrolo[2,3-b]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₅N₃O₂
MDL Number: MFCD10568289
CAS Number: 1053660-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR