Ethyl-5-acetyl-2-phenylthiazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

059020-1G 059020-5G
101916-726EA 436.92 USD
101916-726 101916-728
Ethyl-5-acetyl-2-phenylthiazole-4-carboxylate ≥95%
Ethyl-5-acetyl-2-phenylthiazole-4-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₃NO₃S
MDL Number: MFCD00173944
CAS Number: 57560-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR