Ethyl-5-acetyl-2-methylthiazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

059019-1G 059019-5G
101916-722EA 369.23 USD
101916-722 101916-724
Ethyl-5-acetyl-2-methylthiazole-4-carboxylate ≥95%
Ethyl-5-acetyl-2-methylthiazole-4-carboxylate

Hazard: Irritant

Formula: C₉H₁₁NO₃S
MW: 213.26 g/mol
MDL Number: MFCD00173821
CAS Number: 175277-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR