Ethyl-4-acetyl-5-methylisoxazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

059018-5G 059018-10G
101916-718EA 498.46 USD
101916-718 101916-720
Ethyl-4-acetyl-5-methylisoxazole-3-carboxylate ≥95%
Ethyl-4-acetyl-5-methylisoxazole-3-carboxylate

Hazard: Irritant

Formula: C₉H₁₁NO₄
MW: 197.19 g/mol
MDL Number: MFCD00173953
CAS Number: 15911-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR