(E)-Cyclooct-1-enecarbaldehyde, Technical Grade

Supplier: Matrix Scientific

059015-1G 059015-5G
101916-706EA 553.85 USD
101916-706 101916-708
(E)-Cyclooct-1-enecarbaldehyde, Technical Grade
(E)-Cyclooct-1-enecarbaldehyde

Hazard: Irritant

Formula: C₉H₁₄O
MDL Number: MFCD15142793
CAS Number: 96308-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR