(E)-2-Cyano-3-phenylbut-2-enoic acid ethyl ester ≥95%

Supplier: Matrix Scientific

059014-1G 059014-5G 059014-10G
101916-700EA 175.38 USD
101916-700 101916-702 101916-704
(E)-2-Cyano-3-phenylbut-2-enoic acid ethyl ester ≥95%
(E)-2-Cyano-3-phenylbut-2-enoic acid ethyl ester

Hazard: Irritant

Formula: C₁₃H₁₃NO₂
MDL Number: MFCD00182513
CAS Number: 18300-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR