(E)-4-(3-Bromophenyl)but-3-en-2-one ≥95%

Supplier: Matrix Scientific

059012-1G 059012-5G
101916-690EA 436.92 USD
101916-690 101916-692
(E)-4-(3-Bromophenyl)but-3-en-2-one ≥95%
(E)-4-(3-Bromophenyl)but-3-en-2-one

Hazard: Irritant

Formula: C₁₀H₉BrO
MDL Number: MFCD11845757
CAS Number: 65300-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR