4-[3-(1,3-Dioxo-1,3-dihydroisoindol-2-yl)propyl]piperazine-1-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

059009-1G 059009-5G
101916-682EA 553.85 USD
101916-682 101916-684
4-[3-(1,3-Dioxo-1,3-dihydroisoindol-2-yl)propyl]piperazine-1-carboxylic acid tert-butyl ester ≥95%
4-[3-(1,3-Dioxo-1,3-dihydroisoindol-2-yl)propyl]piperazine-1-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₂₀H₂₇N₃O₄
MW: 373.46 g/mol
MDL Number: MFCD09029311
CAS Number: 857266-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR