Dimethyl(5-trifluoromethylpyridin-2-yl)amine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Dimethylamino-5-(trifluoromethyl)pyridine

059008-1G 059008-5G
101916-678EA 553.85 USD
101916-678 101916-680
Dimethyl(5-trifluoromethylpyridin-2-yl)amine ≥95%
Dimethyl(5-trifluoromethylpyridin-2-yl)amine

Hazard: Irritant

Formula: C₈H₉F₃N₂
MW: 190.17 g/mol
MDL Number: MFCD00766812
CAS Number: 136539-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR