1,3-Dimethyl-1H-pyrazole-5-carbonyl chloride ≥95%

Supplier: Matrix Scientific

059003-1G 059003-5G
101916-662EA 263.08 USD
101916-662 101916-664
1,3-Dimethyl-1H-pyrazole-5-carbonyl chloride ≥95%
1,3-Dimethyl-1H-pyrazole-5-carbonyl chloride

Hazard: Irritant

Formula: C₆H₇ClN₂O
MDL Number: MFCD00833258
CAS Number: 55458-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR