Ethyl trimethylacetopyruvate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5,5-dimethyl-2,4-dioxohexanoate

059001-1G 059001-10G
101916-656EA 116.92 USD
101916-656 101916-658
Ethyl trimethylacetopyruvate ≥95%
Ethyl trimethylacetopyruvate

Hazard: Irritant

Formula: C₁₀H₁₆O₄
MW: 200.24
Boiling Pt: 68…70 °C (0.5 mmHg)
MDL Number: MFCD00052319
CAS Number: 13395-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR