N-[1-Dimethylaminomethylidene]benzamide ≥95%

Supplier: Matrix Scientific

058992-1G 058992-5G
101916-624EA 184.62 USD
101916-624 101916-626
N-[1-Dimethylaminomethylidene]benzamide ≥95%
N-[1-Dimethylaminomethylidene]benzamide

Hazard: Irritant

Formula: C₁₀H₁₂N₂O
MDL Number: MFCD00187256
CAS Number: 41876-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR