(R)-2-(2,5-Dimethoxyphenyl)thiazolidine-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

058990-1G 058990-5G 058990-10G
101916-616EA 263.08 USD
101916-616 101916-618 101916-620
(R)-2-(2,5-Dimethoxyphenyl)thiazolidine-4-carboxylic acid ≥95%
(R)-2-(2,5-Dimethoxyphenyl)thiazolidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₅NO₄S
MDL Number: MFCD10568299
CAS Number: 637032-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR