5-Dimethoxymethyl-1-phenyl-1H-pyrazole-4-carboxylic acid methyl ester ≥95%

Supplier: Matrix Scientific

058988-1G 058988-5G
101916-610EA 436.92 USD
101916-610 101916-612
5-Dimethoxymethyl-1-phenyl-1H-pyrazole-4-carboxylic acid methyl ester ≥95%
5-Dimethoxymethyl-1-phenyl-1H-pyrazole-4-carboxylic acid methyl ester

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₄
MDL Number: MFCD02090518
CAS Number: 957947-49-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR