3,4-Dimethoxybenzamidoxime ≥95%

Supplier: Matrix Scientific

058985-1G 058985-5G 058985-10G
101916-594EA 263.08 USD
101916-594 101916-596 101916-598
3,4-Dimethoxybenzamidoxime ≥95%
3,4-Dimethoxybenzamidoxime

Hazard: Irritant

Formula: C₉H₁₂N₂O₃
MDL Number: MFCD06147747
CAS Number: 40312-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR