3,4-Dihydro-2H-benzo[1,4]oxazine-2-carboxylicacid ethyl ester hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl benzomorpholine-2-carboxylate hydrochloride

058983-1G 058983-5G 058983-10G
101916-584EA 378.46 USD
101916-584 101916-586 101916-588
3,4-Dihydro-2H-benzo[1,4]oxazine-2-carboxylicacid ethyl ester hydrochloride ≥95%
3,4-Dihydro-2H-benzo[1,4]oxazine-2-carboxylicacid ethyl ester hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄ClNO₃
MW: 243.69 g/mol
MDL Number: MFCD11227230
CAS Number: 13582-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR