N-(2,4-Difluorophenyl)pyrrolidin-2-one-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

058979-1G 058979-5G
101916-572EA 553.85 USD
101916-572 101916-574
N-(2,4-Difluorophenyl)pyrrolidin-2-one-4-carboxylic acid ≥95%
N-(2,4-Difluorophenyl)pyrrolidin-2-one-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₉F₂NO₃
MDL Number: MFCD08444772
CAS Number: 923178-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR