2,4-Difluorobenzamidine hydrochloride ≥95%

Supplier: Matrix Scientific

058976-1G 058976-5G 058976-10G
101916-556EA 152.31 USD
101916-556 101916-558 101916-560
2,4-Difluorobenzamidine hydrochloride ≥95%
2,4-Difluorobenzamidine hydrochloride

Hazard: Irritant

Formula: C₇H₇ClF₂N₂
Melting Pt: 100…103 °C
MDL Number: MFCD04114445
CAS Number: 885957-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR