(3,4-Dichlorophenyl)morpholin-4-ylacetonitrile hydrochloride ≥95%

Supplier: Matrix Scientific

058974-1G 058974-5G 058974-10G
101916-548EA 204.62 USD
101916-548 101916-550 101916-552
(3,4-Dichlorophenyl)morpholin-4-ylacetonitrile hydrochloride ≥95%
(3,4-Dichlorophenyl)morpholin-4-ylacetonitrile hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₃Cl₃N₂O
MDL Number: MFCD11052444
CAS Number: 1208081-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR