4,6-Dichloro-2-(methylthio)-5-nitropyrimidine ≥95%

Supplier: Matrix Scientific

058972-1G 058972-5G
101916-540EA 292.31 USD
101916-540 101916-542
4,6-Dichloro-2-(methylthio)-5-nitropyrimidine ≥95%
4,6-Dichloro-2-(methylthio)-5-nitropyrimidine

Hazard: Irritant

Formula: C₅H₃Cl₂N₃O₂S
MW: 240.0672 g/mol
MDL Number: MFCD02323208
CAS Number: 1979-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR