2,6-Dichloroisonicotinoyl chloride ≥95%

Supplier: Matrix Scientific

Synonyms: 2,6-Dichloropyridine-4-carbonyl chloride

058970-1G 058970-5G
101916-530EA 106.15 USD
101916-530 101916-532
2,6-Dichloroisonicotinoyl chloride ≥95%
2,6-Dichloroisonicotinoyl chloride

Hazard: Irritant

Formula: C₆H₂Cl₃NO
MW: 210.45 g/mol
MDL Number: MFCD00052639
CAS Number: 42521-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR