2-(2,6-Dichlorobenzylsulfanyl)ethylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Aminoethyl 2,6-dichlorobenzyl sulphide

058967-1G 058967-5G
101916-522EA 263.08 USD
101916-522 101916-524
2-(2,6-Dichlorobenzylsulfanyl)ethylamine ≥95%
2-(2,6-Dichlorobenzylsulfanyl)ethylamine

Hazard: Irritant

Formula: C₉H₁₁Cl₂NS
MW: 236.16 g/mol
MDL Number: MFCD00052684
CAS Number: 48133-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR