3,4-Dichloro-α-chlorobenzaldoxime, Technical Grade

Supplier: Matrix Scientific

058964-500MG 058964-1G 058964-5G
101916-510EA 466.15 USD
101916-510 101916-512 101916-514
3,4-Dichloro-α-chlorobenzaldoxime, Technical Grade
3,4-Dichloro-alpha-chlorobenzaldoxime

Hazard: Irritant

Formula: C₇H₄Cl₃NO
MDL Number: MFCD11227171
CAS Number: 1056189-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR