3,5-Dibromo-4-pyridone ≥95%

Supplier: Matrix Scientific

058963-1G 058963-5G
101916-506EA 23.08 USD
101916-506 101916-508
3,5-Dibromo-4-pyridone ≥95%
3,5-Dibromo-4-pyridone

Hazard: Irritant

Formula: C₅H₃Br₂NO
MDL Number: MFCD00082969
CAS Number: 141375-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR