2,3-Dibromo-propionitrile ≥95%

Supplier: Matrix Scientific

058962-25G 058962-5G
101916-504EA 707.81 USD
101916-504 102833-272
2,3-Dibromo-propionitrile ≥95%
2,3-Dibromo-propionitrile

Hazard: Irritant

Formula: C₃H₃Br₂N
Boiling Pt: 67…70 °C (0.5 mmHg)
MDL Number: MFCD00001857
CAS Number: 4554-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR