2,2-Dibromo-3-oxobutyric acid ethyl ester ≥95%

Supplier: Matrix Scientific

058960-1G 058960-5G
101916-496EA 436.92 USD
101916-496 101916-498
2,2-Dibromo-3-oxobutyric acid ethyl ester ≥95%
2,2-Dibromo-3-oxobutyric acid ethyl ester

Hazard: Irritant

Formula: C₆H₈Br₂O₃
MW: 287.95 g/mol
MDL Number: MFCD11845706
CAS Number: 89415-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR