5,7-Dibromo-1,3-dihydroindol-2-one ≥95%

Supplier: Matrix Scientific

058959-1G 058959-5G 058959-10G
101916-490EA 200 USD
101916-490 101916-492 101916-494
5,7-Dibromo-1,3-dihydroindol-2-one ≥95%
5,7-Dibromo-1,3-dihydroindol-2-one

Hazard: Irritant

Formula: C₈H₅Br₂NO
MDL Number: MFCD04230619
CAS Number: 23872-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR