3,3-Dibromoazepan-2-one ≥95%

Supplier: Matrix Scientific

058958-500MG 058958-1G 058958-5G
101916-484EA 466.15 USD
101916-484 101916-486 101916-488
3,3-Dibromoazepan-2-one ≥95%
3,3-Dibromoazepan-2-one

Hazard: Irritant

Formula: C₆H₉Br₂NO
MDL Number: MFCD00154706
CAS Number: 56987-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR