N'-[1-Cyclopropylaminopropylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%

Supplier: Matrix Scientific

058957-500MG 058957-1G
101916-480EA 284.62 USD
101916-480 101916-482
N'-[1-Cyclopropylaminopropylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Cyclopropylaminopropylidene]hydrazinecarboxylic acid-tert-butyl ester

Hazard: Irritant

Formula: C₁₁H₂₁N₃O₂
MW: 227.31 g/mol
MDL Number: MFCD10568246
CAS Number: 1053657-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR