N'-[1-Cyclopropylamino-2-phenylethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%

Supplier: Matrix Scientific

058956-500MG 058956-1G
101916-476EA 284.62 USD
101916-476 101916-478
N'-[1-Cyclopropylamino-2-phenylethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Cyclopropylamino-2-phenylethylidene]hydrazinecarboxylic acid-tert-butyl ester

Hazard: Irritant

Formula: C₁₆H₂₃N₃O₂
MW: 289.38 g/mol
MDL Number: MFCD10568248
CAS Number: 1053657-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR