N'-[1-Cyclopropylamino-2-phenoxyethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%

Supplier: Matrix Scientific

058955-500MG 058955-1G
101916-472EA 284.62 USD
101916-472 101916-474
N'-[1-Cyclopropylamino-2-phenoxyethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Cyclopropylamino-2-phenoxyethylidene]hydrazinecarboxylic acid-tert-butyl ester

Hazard: Irritant

Formula: C₁₆H₂₃N₃O₃
MW: 305.38 g/mol
MDL Number: MFCD10568249
CAS Number: 1053657-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR