N'-[1-Cyclopentylamino-2-phenylethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%

Supplier: Matrix Scientific

058952-500MG 058952-1G
101916-458EA 284.62 USD
101916-458 101916-460
N'-[1-Cyclopentylamino-2-phenylethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Cyclopentylamino-2-phenylethylidene]hydrazinecarboxylic acid-tert-butyl ester

Hazard: Irritant

Formula: C₁₈H₂₇N₃O₂
MW: 317.43 g/mol
MDL Number: MFCD10568256
CAS Number: 1053657-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR