N'-[1-Cyclopentylamino-2-(4-methoxyphenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%

Supplier: Matrix Scientific

058950-500MG 058950-1G
101916-450EA 284.62 USD
101916-450 101916-452
N'-[1-Cyclopentylamino-2-(4-methoxyphenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Cyclopentylamino-2-(4-methoxyphenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester

Hazard: Irritant

Formula: C₁₉H₂₉N₃O₃
MW: 347.46 g/mol
MDL Number: MFCD10568254
CAS Number: 1053657-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR