N'-[1-Cyclobutylaminopropylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%

Supplier: Matrix Scientific

058949-500MG 058949-1G
101916-446EA 284.62 USD
101916-446 101916-448
N'-[1-Cyclobutylaminopropylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Cyclobutylaminopropylidene]hydrazinecarboxylic acid-tert-butyl ester

Hazard: Irritant

Formula: C₁₂H₂₃N₃O₂
MW: 241.34 g/mol
MDL Number: MFCD10568258
CAS Number: 1053657-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR