N'-[1-Cyclobutylamino-2-phenylethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%

Supplier: Matrix Scientific

058948-500MG 058948-1G
101916-442EA 284.62 USD
101916-442 101916-444
N'-[1-Cyclobutylamino-2-phenylethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Cyclobutylamino-2-phenylethylidene]hydrazinecarboxylic acid-tert-butyl ester

Hazard: Irritant

Formula: C₁₇H₂₅N₃O₂
MW: 303.41 g/mol
MDL Number: MFCD10568257
CAS Number: 1053657-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR