N'-[1-Cyclobutylamino-2-phenoxyethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%

Supplier: Matrix Scientific

058947-500MG 058947-1G
101916-438EA 284.62 USD
101916-438 101916-440
N'-[1-Cyclobutylamino-2-phenoxyethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Cyclobutylamino-2-phenoxyethylidene]hydrazinecarboxylic acid-tert-butyl ester

Hazard: Irritant

Formula: C₁₇H₂₅N₃O₃
MW: 319.41 g/mol
MDL Number: MFCD10568244
CAS Number: 1053657-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR