4-(3-Cyanopyridin-2-yl)benzoic acid ethyl ester ≥95%

Supplier: Matrix Scientific

058946-500MG 058946-1G
101916-434EA 549.23 USD
101916-434 101916-436
4-(3-Cyanopyridin-2-yl)benzoic acid ethyl ester ≥95%
4-(3-Cyanopyridin-2-yl)benzoic acid ethyl ester

Hazard: Irritant

Formula: C₁₅H₁₂N₂O₂
Melting Pt: 98…100 °C
MDL Number: MFCD15142822
CAS Number: 1208081-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR