1-(Cyanophenylmethyl)piperidine-4-carboxylicacid ethyl ester hydrochloride ≥95%

Supplier: Matrix Scientific

058944-1G 058944-5G 058944-10G
101916-426EA 204.62 USD
101916-426 101916-428 101916-430
1-(Cyanophenylmethyl)piperidine-4-carboxylicacid ethyl ester hydrochloride ≥95%
1-(Cyanophenylmethyl)piperidine-4-carboxylicacid ethyl ester hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₁ClN₂O₂
MDL Number: MFCD11052433
CAS Number: 1208081-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR