3-Cyano-2-mercapto-6-methylisonicotinic acid ethyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-cyano-6-methyl-2-sulphanylisonicotinate

058940-1G 058940-5G 058940-10G
101916-406EA 175.38 USD
101916-406 101916-408 101916-410
3-Cyano-2-mercapto-6-methylisonicotinic acid ethyl ester ≥95%
3-Cyano-2-mercapto-6-methylisonicotinic acid ethyl ester

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₂S
MW: 222.27 g/mol
MDL Number: MFCD00052275
CAS Number: 56891-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR