N-[3-Cyano-4,5-dimethyl-1-(2,4,6-trimethylphenyl)-1H-pyrrol-2-yl]acetamide ≥95%

Supplier: Matrix Scientific

058939-1G 058939-5G
101916-402EA 495.38 USD
101916-402 101916-404
N-[3-Cyano-4,5-dimethyl-1-(2,4,6-trimethylphenyl)-1H-pyrrol-2-yl]acetamide ≥95%
N-[3-Cyano-4,5-dimethyl-1-(2,4,6-trimethylphenyl)-1H-pyrrol-2-yl]acetamide

Hazard: Irritant

Formula: C₁₈H₂₁N₃O
MW: 295.39 g/mol
MDL Number: MFCD18384850
CAS Number: 157286-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR