2-Cyano-5-bromomethylpyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(Bromomethyl)picolinonitrile, 5-(Bromomethyl)pyridine-2-carbonitrile

058938-250MG 058938-1G
101916-398EA 186.15 USD
101916-398 101916-400
2-Cyano-5-bromomethylpyridine ≥95%
2-Cyano-5-bromomethylpyridine

Hazard: Irritant

Formula: C₇H₅BrN₂
MW: 197.04 g/mol
MDL Number: MFCD09037857
CAS Number: 308846-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR