1'-(6-Chloro-4-(trifluoromethyl)pyridin-2-yl)piperazine ≥95%

Supplier: Matrix Scientific

058935-1G 058935-5G
101916-386EA 307.69 USD
101916-386 101916-388
1'-(6-Chloro-4-(trifluoromethyl)pyridin-2-yl)piperazine ≥95%
1'-(6-Chloro-4-(trifluoromethyl)pyridin-2-yl)piperazine

Hazard: Irritant

Formula: C₁₀H₁₁ClF₃N₃
MDL Number: MFCD09991799
CAS Number: 750628-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR