[3-Chloro-5-trifluoromethyl-1H-pyridin-(2E)-ylidene]cyanoacetic acid ethyl ester ≥95%

Supplier: Matrix Scientific

058933-1G 058933-5G
101916-380EA 553.85 USD
101916-380 101916-382
[3-Chloro-5-trifluoromethyl-1H-pyridin-(2E)-ylidene]cyanoacetic acid ethyl ester ≥95%
[3-Chloro-5-trifluoromethyl-1H-pyridin-(2E)-ylidene]cyanoacetic acid ethyl ester

Hazard: Irritant

Formula: C₁₁H₈ClF₃N₂O₂
MW: 292.65 g/mol
MDL Number: MFCD18384848
CAS Number: 1300019-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR