(6-Chloro-4-(trifluoromethyl)pyridin-2-yl)methylamine ≥95%
Supplier: Matrix Scientific
058931-500MG
058931-1G
101916-374EA
553.85
USD
101916-374
101916-376
(6-Chloro-4-(trifluoromethyl)pyridin-2-yl)methylamine ≥95%
(6-Chloro-4-(trifluoromethyl)pyridin-2-yl)methylamine
Hazard: Irritant
Formula:
C₇H₆ClF₃N₂ |
MDL Number:
MFCD15142767 CAS Number: 1208081-32-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...